siRNA Details
Gene Symbol: Syt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172804.2 | NP_766392.2 | 4 | 555 | ||
GenBank mRNA | AB102950.1 | - | 3 | 887 | ||
AK032606.1 | - | 5 | 1333 | |||
AK137129.1 | - | 4 | 563 | |||
BC056932.1 | - | 3 | 665 | 5708126 |
