siRNA Details
| Gene Symbol: Adam6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009545.1 | NP_001009545.1 | 1 | 2252 | ||
| GenBank mRNA | AK076580.1 | - | 1 | 2308 | ||
| BC111877.1 | - | 1 | 2272 | 40059665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam6b| Gene Symbol: Adam6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009545.1 | NP_001009545.1 | 1 | 2252 | ||
| GenBank mRNA | AK076580.1 | - | 1 | 2308 | ||
| BC111877.1 | - | 1 | 2272 | 40059665 | ||