siRNA Details
| Gene Symbol: Tubal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033879.3 | NP_001029051.2 | 4 | 650 | ||
| GenBank mRNA | AK135971.1 | - | 4 | 651 | ||
| BC147507.1 | - | 4 | 643 | 9056442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tubal3| Gene Symbol: Tubal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033879.3 | NP_001029051.2 | 4 | 650 | ||
| GenBank mRNA | AK135971.1 | - | 4 | 651 | ||
| BC147507.1 | - | 4 | 643 | 9056442 | ||