siRNA Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243046.1 | NP_001229975.1 | 15 | 1559 | ||
| NM_001243047.1 | NP_001229976.1 | 11 | 1242 | |||
| NM_011807.3 | NP_035937.2 | 23 | 2718 | |||
| GenBank mRNA | AF388675.1 | - | 23 | 2497 | ||
| AK030221.1 | - | 15 | 1561 | |||
| AK039754.1 | - | 11 | 1107 | |||

Dlg2
Genomic Map