siRNA Details
| Gene Symbol: Spata31d1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167593.1 | NP_001161065.1 | 4 | 1394 | ||
| GenBank mRNA | AK133300.1 | - | 4 | 1396 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata31d1b| Gene Symbol: Spata31d1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167593.1 | NP_001161065.1 | 4 | 1394 | ||
| GenBank mRNA | AK133300.1 | - | 4 | 1396 | ||