siRNA Details
| Gene Symbol: Gpr150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175495.2 | NP_780704.1 | 1 | 322 | ||
| GenBank mRNA | AK047609.1 | - | 1 | 322 | ||
| AY255541.1 | - | 1 | 126 | |||
| BC118612.1 | - | 1 | 302 | 40107174 | ||
| BC118614.1 | - | 1 | 303 | 40107172 | ||
More Information
Set Membership: |
|

Gpr150
Genomic Map