siRNA Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011812.4 | NP_035942.1 | 4 | 611 | ||
| GenBank mRNA | AF112151.1 | - | 4 | 561 | ||
| AK084895.1 | - | 4 | 452 | |||
| AK085170.1 | - | 4 | 441 | |||
| AK090129.1 | - | 4 | 550 | |||
| AK159189.1 | - | 4 | 374 | |||
| AK159471.1 | - | 4 | 594 | |||
| AK159960.1 | - | 4 | 581 | |||
| AK165018.1 | - | 4 | 404 | |||
| BC006636.1 | - | 4 | 301 | 3482574 | ||

Fbln5
Genomic Map