siRNA Details
| Gene Symbol: Pde4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011056.3 | NP_035186.1 | 4 | 628 | ||
| GenBank mRNA | AF038896.1 | - | 2 | 310 | ||
| AF536978.1 | - | 4 | 628 | |||
| AK039354.1 | - | 2 | 222 | |||
| AK039692.1 | - | 2 | 222 | |||
| AK081013.1 | - | 3 | 929 | |||
| AY388962.1 | - | 2 | 241 | |||
| BC132107.1 | - | 3 | 496 | 40130384 | ||
| BC132109.1 | - | 3 | 496 | 40130386 | ||
| EU489880.1 | - | 2 | 484 | |||

Pde4d
Genomic Map