siRNA Details
| Gene Symbol: Grem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011825.1 | NP_035955.1 | 2 | 383 | ||
| GenBank mRNA | AB011030.1 | - | 2 | 383 | ||
| AK142008.1 | - | 2 | 385 | |||
| AK161827.1 | - | 2 | 363 | |||
| BC079905.1 | - | 2 | 364 | 30645267 | ||

Grem2
Genomic Map