siRNA Details
| Gene Symbol: Rhof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175092.3 | NP_780301.1 | 3 | 545 | ||
| GenBank mRNA | AK038837.1 | - | 313 | |||
| AK172212.1 | - | 3 | 313 | |||
| BC096597.1 | - | 3 | 335 | 4983233 | ||
| BC132355.1 | - | 377 | 40130632 | |||
| BC132357.1 | - | 377 | 40130634 | |||
| CK617811.1 | - | 3 | 552 | |||

Rhof
Genomic Map