siRNA Details
Gene Symbol: Rhof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175092.3 | NP_780301.1 | 3 | 578 | ||
GenBank mRNA | AK038837.1 | - | 346 | |||
AK172212.1 | - | 3 | 346 | |||
BC096597.1 | - | 3 | 368 | 4983233 | ||
BC132355.1 | - | 410 | 40130632 | |||
BC132357.1 | - | 410 | 40130634 | |||
CK617811.1 | - | 3 | 585 |
