siRNA Details
| Gene Symbol: Gucy1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204340.1 | NP_001191269.1 | 17 | 2427 | ||
| NM_172810.3 | NP_766398.1 | 17 | 2472 | |||
| GenBank mRNA | AK047129.1 | - | 17 | 2473 | ||
| AY236513.1 | - | 15 | 2146 | |||
| BC137868.1 | - | 17 | 2424 | 8860887 | ||
| BC144988.1 | - | 17 | 2421 | 9053536 | ||
| BY391897.1 | - | 2 | 166 | |||

Gucy1b2
Genomic Map