siRNA Details
| Gene Symbol: Insl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290648.1 | NP_001277577.1 | 3 | 369 | ||
| NM_011831.2 | NP_035961.1 | 2 | 375 | |||
| GenBank mRNA | AF054842.1 | - | 3 | 369 | ||
| AF054843.1 | - | 2 | 339 | |||
| AF076971.1 | - | 2 | 371 | |||
| AF133817.1 | - | 2 | 378 | |||
| BC010968.1 | - | 2 | 370 | 4217383 | ||

Insl5
Genomic Map