siRNA Details
| Gene Symbol: Krtap14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013707.2 | NP_038735.2 | 1 | 114 | ||
| GenBank mRNA | AF003691.1 | - | 1 | 106 | ||
| BC104262.1 | - | 1 | 88 | 40040069 | ||
| BC104263.1 | - | 1 | 87 | 40040071 | ||
| CJ056567.1 | - | 1 | 115 | |||
| D85925.1 | - | 1 | 103 | |||

Krtap14
Genomic Map