siRNA Details
| Gene Symbol: Aard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175503.3 | NP_780712.2 | 1 | 214 | ||
| GenBank mRNA | AK046414.1 | - | 1 | 215 | ||
| AY134665.1 | - | 1 | 153 | |||
| BC089505.1 | - | 1 | 168 | 6772439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aard| Gene Symbol: Aard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175503.3 | NP_780712.2 | 1 | 214 | ||
| GenBank mRNA | AK046414.1 | - | 1 | 215 | ||
| AY134665.1 | - | 1 | 153 | |||
| BC089505.1 | - | 1 | 168 | 6772439 | ||