siRNA Details
| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011846.4 | NP_035976.3 | 9 | 1415 | ||
| GenBank mRNA | AB021224.2 | - | 9 | 1421 | ||
| AJ010731.1 | - | 9 | 1419 | |||
| AK079221.1 | - | 1187 | ||||
| AK143521.1 | - | 9 | 1415 | |||
| AK179012.1 | - | 1 | 45 | |||
| BC051917.1 | - | 9 | 1415 | 5716458 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001105925, XM_008769195.

Mmp17
Genomic Map