siRNA Details
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153178.4 | NP_694818.3 | 15 | 2092 | ||
GenBank mRNA | AB081472.1 | - | 15 | 1954 | ||
AK220193.1 | - | 12 | 1457 | |||
BC023279.1 | - | 7 | 793 | |||
BC024857.1 | - | 8 | 937 | |||
BC056639.1 | - | 5 | 659 | |||
BC064741.1 | - | 10 | 1209 | |||
BC096465.1 | - | 15 | 1997 | |||
BC128379.1 | - | 15 | 1945 | 40111876 | ||
BC129922.1 | - | 15 | 1945 | 40111875 |
