siRNA Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178719.5 | NP_848834.2 | 6 | 1748 | ||
| GenBank mRNA | AK038466.1 | - | 6 | 1721 | ||
| AK041184.1 | - | 6 | 1641 | |||
| AK044773.1 | - | 7 | 2181 | |||
| AK050546.1 | - | 6 | 1709 | |||
| AK080375.1 | - | 6 | 1694 | |||
| AK163389.1 | - | 7 | 1850 | |||
| BC094597.1 | - | 6 | 1678 | |||
| BC137633.1 | - | 5 | 1652 | 8860652 | ||

Mief1
Genomic Map