siRNA Details
| Gene Symbol: A4galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004150.3 | NP_001004150.1 | 3 | 1173 | ||
| NM_001170954.1 | NP_001164425.1 | 3 | 1213 | |||
| GenBank mRNA | AK139833.1 | - | 3 | 1177 | ||
| AK149186.1 | - | 3 | 1129 | |||
| AK162529.1 | - | 3 | 1212 | |||
| AY371179.1 | - | 3 | 1132 | |||
| BC138844.1 | - | 2 | 1088 | 8861866 | ||
| BC138845.1 | - | 2 | 1088 | 8861867 | ||
| DQ143885.1 | - | 1 | 907 | |||

A4galt
Genomic Map