siRNA Details
| Gene Symbol: Ttll8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172818.3 | NP_766406.2 | 8 | 1396 | ||
| GenBank mRNA | AK029745.1 | - | 8 | 1392 | ||
| AK030151.1 | - | 8 | 1389 | |||
| AM690752.1 | - | 8 | 1053 | |||
| BC116293.1 | - | 8 | 1037 | 40094952 | ||
| BC116294.1 | - | 8 | 1032 | 40094955 | ||

Ttll8
Genomic Map