siRNA Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122667.2 | NP_001116139.1 | 5 | 414 | ||
| NM_153588.3 | NP_705816.2 | 3 | 428 | |||
| GenBank mRNA | AF532598.1 | - | 3 | 210 | ||
| AK220377.1 | - | 3 | 210 | |||
| BC131644.1 | - | 3 | 210 | 40110019 | ||
| BC131645.1 | - | 3 | 208 | |||
| CJ126058.1 | - | 3 | 430 | |||

Mkl2
Genomic Map