siRNA Details
| Gene Symbol: Rtp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004151.2 | NP_001004151.1 | 1 | 292 | ||
| GenBank mRNA | AK047543.1 | - | 1 | 293 | ||
| AY562225.1 | - | 1 | 246 | |||
| BC138910.1 | - | 1 | 278 | 8861932 | ||
| BC138912.1 | - | 1 | 278 | 8861934 | ||
| EU070411.1 | - | 1 | 138 | |||

Rtp1
Genomic Map