siRNA Details
| Gene Symbol: Prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011871.2 | NP_036001.1 | 5 | 627 | ||
| GenBank mRNA | AF083032.1 | - | 5 | 627 | ||
| AK012798.1 | - | 5 | 613 | |||
| AK155112.1 | - | 5 | 651 | |||
| BC011311.1 | - | 5 | 635 | 3985758 | ||

Prkra
Genomic Map