siRNA Details
Gene Symbol: Zfp799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177359.5 | NP_796333.3 | 4 | 673 | ||
GenBank mRNA | AK031686.1 | - | 4 | 661 | ||
AK083640.1 | - | 4 | 650 | |||
AK137037.1 | - | 4 | 650 | |||
BC096552.1 | - | 4 | 623 | 30618827 |
