siRNA Details
| Gene Symbol: Ythdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163013.1 | NP_001156485.1 | 26 | 3815 | ||
| GenBank mRNA | BC037178.1 | - | 13 | 1811 | ||
| BC040772.1 | - | 7 | 1102 | |||
| BC072654.1 | - | 21 | 2859 | |||
| BC138263.1 | - | 26 | 3815 | 8861282 | ||
| BC171951.1 | - | 26 | 3815 | 9054237 | ||

Ythdc2
Genomic Map