siRNA Details
| Gene Symbol: Loxhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172834.2 | NP_766422.2 | 2, 3 | 250 | ||
| GenBank mRNA | FJ750876.1 | - | 2, 3 | 250 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Loxhd1| Gene Symbol: Loxhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172834.2 | NP_766422.2 | 2, 3 | 250 | ||
| GenBank mRNA | FJ750876.1 | - | 2, 3 | 250 | ||