siRNA Details
| Gene Symbol: Adnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175028.1 | NP_778193.1 | 4 | 2887 | ||
| GenBank mRNA | AB093268.1 | - | 4 | 2887 | ||
| BC024969.1 | - | 1 | 124 | |||
| BC044898.1 | - | 4 | 2792 | |||
| BC044904.1 | - | 1 | 238 | |||
| BC047352.1 | - | 4 | 2792 | |||
| BC064672.1 | - | 2 | 2660 | |||

Adnp2
Genomic Map