siRNA Details
| Gene Symbol: Sgcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011890.4 | NP_036020.1 | 5 | 758 | ||
| GenBank mRNA | AB024921.1 | - | 5 | 772 | ||
| AK014381.2 | - | 5 | 772 | |||
| AK017781.1 | - | 5 | 762 | |||
| AK132306.1 | - | 5 | 817 | |||
| AK169403.1 | - | 5 | 759 | |||
| BC052349.1 | - | 5 | 748 | 6744003 | ||

Sgcb
Genomic Map