siRNA Details
Gene Symbol: Sgcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011890.4 | NP_036020.1 | 6 | 912 | ||
GenBank mRNA | AB024921.1 | - | 6 | 926 | ||
AK014381.2 | - | 6 | 926 | |||
AK017781.1 | - | 6 | 916 | |||
AK132306.1 | - | 6 | 971 | |||
AK169403.1 | - | 6 | 913 | |||
BC052349.1 | - | 6 | 902 | 6744003 |
