siRNA Details
| Gene Symbol: Spry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011898.2 | NP_036028.2 | 2 | 727 | ||
| GenBank mRNA | AB019280.1 | - | 2 | 796 | ||
| AF176906.1 | - | 2 | 880 | |||
| AK037180.1 | - | 2 | 802 | |||
| AK048122.1 | - | 2 | 805 | |||
| BC050944.1 | - | 2 | 727 | 6821944 | ||
| BC057005.1 | - | 2 | 727 | 6821944 | ||

Spry4
Genomic Map