siRNA Details
| Gene Symbol: Teddm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178244.3 | NP_839975.2 | 1 | 242 | ||
| GenBank mRNA | AJ554212.1 | - | 1 | 96 | ||
| BC109332.2 | - | 1 | 95 | |||
| BC141344.1 | - | 1 | 145 | 9055956 | ||
| BC141345.1 | - | 1 | 146 | 9055957 | ||
| CJ048231.1 | - | 1 | 408 | |||

Teddm1a
Genomic Map