siRNA Details
| Gene Symbol: Fam78a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175511.4 | NP_780720.1 | 1 | 1050 | ||
| GenBank mRNA | AK077626.1 | - | 1 | 1055 | ||
| AK079514.1 | - | 1 | 1051 | |||
| AK165375.1 | - | 1418 | ||||
| BC137951.1 | - | 1 | 544 | 8860970 | ||
| BC137952.1 | - | 1 | 544 | 8860971 | ||

Fam78a
Genomic Map