siRNA Details
Gene Symbol: Dhrs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175512.2 | NP_780721.1 | 3 | 562 | ||
GenBank mRNA | AF361479.1 | - | 2 | 347 | ||
AK050180.1 | - | 3 | 580 | |||
AK080914.1 | - | 3 | 562 | |||
AK089247.1 | - | 174 | ||||
AY936479.1 | - | 2 | 334 | |||
BC118029.1 | - | 3 | 523 | 40093681 |
