siRNA Details
| Gene Symbol: Lrrc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033346.2 | NP_001028518.1 | 1 | 111 | ||
| GenBank mRNA | AK134968.1 | - | 112 | |||
| BC141221.1 | - | 111 | 9055833 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc55| Gene Symbol: Lrrc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033346.2 | NP_001028518.1 | 1 | 111 | ||
| GenBank mRNA | AK134968.1 | - | 112 | |||
| BC141221.1 | - | 111 | 9055833 | |||