siRNA Details
Gene Symbol: Lrrc55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033346.2 | NP_001028518.1 | 1 | 171 | ||
GenBank mRNA | AK134968.1 | - | 172 | |||
BC141221.1 | - | 171 | 9055833 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrc55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033346.2 | NP_001028518.1 | 1 | 171 | ||
GenBank mRNA | AK134968.1 | - | 172 | |||
BC141221.1 | - | 171 | 9055833 |