siRNA Details
| Gene Symbol: Tspan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183180.2 | NP_899003.1 | 8 | 861 | ||
| GenBank mRNA | AK032765.1 | - | 7 | 772 | ||
| BC052169.1 | - | 8 | 824 | 6335221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan18| Gene Symbol: Tspan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183180.2 | NP_899003.1 | 8 | 861 | ||
| GenBank mRNA | AK032765.1 | - | 7 | 772 | ||
| BC052169.1 | - | 8 | 824 | 6335221 | ||