siRNA Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291027.1 | NP_001277956.1 | 5 | 2100 | ||
| NM_001291028.1 | NP_001277957.1 | 5 | 1821 | |||
| NM_197945.4 | NP_922936.3 | 5 | 2163 | |||
| GenBank mRNA | AK148251.1 | - | 5 | 2062 | ||
| AK161711.1 | - | 6 | 2064 | |||
| BC049892.1 | - | 3 | 1724 | |||
| BC058280.1 | - | 4 | 1745 | 6529102 | ||
| CX242174.1 | - | 1 | 317 | |||

Lzts3
Genomic Map