siRNA Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081338.1 | NP_001074807.1 | 21 | 2518 | ||
| GenBank mRNA | AK049859.1 | - | 20 | 2897 | ||
| AK220350.1 | - | 19 | 2266 | |||
| BC116639.1 | - | 18 | 2968 | 40061282 | ||

L3mbtl1
Genomic Map