siRNA Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081338.1 | NP_001074807.1 | 3 | 368 | ||
| GenBank mRNA | AJ875399.1 | - | 2 | 186 | ||
| AJ875401.1 | - | 2 | 186 | |||
| AK049859.1 | - | 3 | 362 | |||
| AK220350.1 | - | 1 | 116 | |||
| BC116639.1 | - | 3 | 432 | 40061282 | ||

L3mbtl1
Genomic Map