siRNA Details
| Gene Symbol: Tmem8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085508.2 | NP_001078977.1 | 6 | 1199 | ||
| GenBank mRNA | AK029713.1 | - | 6 | 1200 | ||
| BC147569.1 | - | 6 | 1070 | 9056559 | ||
| BC147574.1 | - | 6 | 1070 | 9056564 | ||
| BC151067.1 | - | 5 | 801 | 9087980 | ||
| BC151070.1 | - | 6 | 1070 | 9087983 | ||

Tmem8b
Genomic Map