siRNA Details
| Gene Symbol: Tmem245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175518.5 | NP_780727.3 | 16 | 2404 | ||
| GenBank mRNA | AK052826.1 | - | 16 | 2963 | ||
| CF538713.1 | - | 1 | 117 | |||
| CN457695.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem245| Gene Symbol: Tmem245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175518.5 | NP_780727.3 | 16 | 2404 | ||
| GenBank mRNA | AK052826.1 | - | 16 | 2963 | ||
| CF538713.1 | - | 1 | 117 | |||
| CN457695.1 | - | 1 | 114 | |||