siRNA Details
| Gene Symbol: Gpatch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172876.2 | NP_766464.1 | 5 | 1151 | ||
| GenBank mRNA | AK053051.1 | - | 5 | 1152 | ||
| BC096468.1 | - | 5 | 1134 | 3665952 | ||
| BC121828.1 | - | 5 | 1149 | 40107316 | ||
| BC121829.1 | - | 5 | 1126 | 40107315 | ||

Gpatch3
Genomic Map