siRNA Details
| Gene Symbol: Myom3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085509.2 | NP_001078978.1 | 26 | 3299 | ||
| GenBank mRNA | AK033394.1 | - | 2 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myom3| Gene Symbol: Myom3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085509.2 | NP_001078978.1 | 26 | 3299 | ||
| GenBank mRNA | AK033394.1 | - | 2 | 192 | ||