siRNA Details
| Gene Symbol: Lrrc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162983.1 | NP_001156455.1 | 2 | 1122 | ||
| GenBank mRNA | AK038658.1 | - | 2 | 1086 | ||
| AK045936.1 | - | 2 | 674 | |||
| AK078190.1 | - | 2 | 1097 | |||
| BC129963.1 | - | 2 | 1115 | |||
| BC129964.1 | - | 3 | 1045 | |||

Lrrc38
Genomic Map