siRNA Details
| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159963.1 | NP_001153435.1 | 9 | 1557 | ||
| NM_178729.4 | NP_848844.1 | 9 | 1557 | |||
| GenBank mRNA | AK029957.1 | - | 9 | 1505 | ||
| AK041497.1 | - | 9 | 1529 | |||
| AK082931.1 | - | 9 | 1527 | |||
| AK085100.1 | - | 9 | 1530 | |||
| AK088067.1 | - | 9 | 1524 | |||
| AK150664.1 | - | 9 | 1514 | |||
| AK159744.1 | - | 9 | 1546 | |||
| AK171783.1 | - | 9 | 1417 | |||
| BC023291.1 | - | 7 | 1162 | |||
| BC047214.1 | - | 1470 | 6466511 | |||
| BC113798.1 | - | 9 | 1475 | 40062506 | ||

Fbxl5
Genomic Map