siRNA Details
| Gene Symbol: Hsd17b13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163486.1 | NP_001156958.1 | 4 | 613 | ||
| NM_198030.2 | NP_932147.2 | 4 | 613 | |||
| GenBank mRNA | AI048624.1 | - | 4 | 600 | ||
| AY101768.1 | - | 4 | 540 | |||
| BC019427.1 | - | 4 | 589 | 5132342 | ||
| DV660487.1 | - | 1 | 39 | |||

Hsd17b13
Genomic Map