siRNA Details
| Gene Symbol: C130060K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175524.4 | NP_780733.2 | 6 | 1057 | ||
| GenBank mRNA | AK048439.1 | - | 6 | 967 | ||
| AK051723.1 | - | 6 | 991 | |||
| BC119578.1 | - | 6 | 959 | 40058179 | ||
| BC119579.1 | - | 6 | 958 | 40058180 | ||
More Information
Set Membership: |
|

C130060K24Rik
Genomic Map