siRNA Details
| Gene Symbol: Lrrtm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134743.1 | NP_001128215.1 | 2 | 1668 | ||
| NM_001282102.1 | NP_001269031.1 | 2 | 1668 | |||
| NM_178731.5 | NP_848846.3 | 2 | 1422 | |||
| GenBank mRNA | AK032470.1 | - | 1 | 1318 | ||
| AK038656.1 | - | 2 | 1499 | |||
| AY182031.1 | - | 1 | 1362 | |||
| BC037216.1 | - | 2 | 1590 | 5361681 | ||

Lrrtm4
Genomic Map