siRNA Details
| Gene Symbol: Ccdc142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081266.1 | NP_001074735.1 | 8 | 1941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc142| Gene Symbol: Ccdc142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081266.1 | NP_001074735.1 | 8 | 1941 | ||