siRNA Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173775.3 | NP_776136.2 | 7 | 666 | ||
| GenBank mRNA | AK048790.1 | - | 3 | 679 | ||
| BC047211.1 | - | 5 | 348 | 6392183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc37| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173775.3 | NP_776136.2 | 7 | 666 | ||
| GenBank mRNA | AK048790.1 | - | 3 | 679 | ||
| BC047211.1 | - | 5 | 348 | 6392183 | ||